4-Methoxy-2-nitrobenzyl bromide

APOSOR400484-250MGEA 0 EUR
APOSOR400484-250MG APOSOR400484-1G
4-Methoxy-2-nitrobenzyl bromide
4-Methoxy-2-nitrobenzyl bromide
Formula: C₈H₈BrNO₃
MW: 246,058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05148455
CAS Number: 57559-52-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR