2-(3,5-Dichlorophenyl)cyclopropane-1-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 1-Carboxy-2-(3,5-dichlorophenyl)cyclopropane1-(2-Carboxycycloprop-1-yl)-3,5-dichlorobenzene

APOSOR40319-250MGEA 408 EUR
APOSOR40319-250MG APOSOR40319-1G
2-(3,5-Dichlorophenyl)cyclopropane-1-carboxylic acid
2-(3,5-Dichlorophenyl)cyclopropane-1-carboxylic acid
Formula: C₁₀H₈Cl₂O₂
MW: 231,075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12149922
CAS Number: 1183181-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR