2-Bromo-N-ethylacetamide
|
Warning
|
Synonyms:
N-(Bromoacetyl)ethylamine
, 2-Bromo-N-ethylethanamide
APOSOR40485-1GEA
453
EUR
APOSOR40485-1G
2-Bromo-N-ethylacetamide
2-Bromo-N-ethylacetamide
Formula:
C₄H₈BrNO MW: 166,016 g/mol Boiling Pt: 72…77 °C (0,3 mm Hg) Melting Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD06800147 CAS Number: 5327-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...