8-Chloro-1-iodophenazine
Supplier: Apollo Scientific
|
Warning
|
APOSOR40780-1GEA
87.3
EUR
APOSOR40780-1G
APOSOR40780-5G
APOSOR40780-250MG
8-Chloro-1-iodophenazine
8-Chloro-1-iodophenazine
Formula:
C₁₂H₆ClIN₂ MW: 340,547 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32111675 CAS Number: 2367002-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...