(2-Bromoethyl)cyclopropane

Supplier: Apollo Scientific
Warning

APOSOR41170-1GEA 669 EUR
APOSOR41170-1G
(2-Bromoethyl)cyclopropane
(2-Bromoethyl)cyclopropane
Formula: C₅H₉Br
MW: 149,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16860592
CAS Number: 36982-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR