2-(2,3-Dimethylphenyl)acetonitrile

Warning

APOSOR41183-1GEA 0 EUR
APOSOR41183-1G APOSOR41183-250MG
2-(2,3-Dimethylphenyl)acetonitrile
2-(2,3-Dimethylphenyl)acetonitrile
Formula: C₁₀H₁₁N
MW: 145,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834792
CAS Number: 76574-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR