3-(2-Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, pinacol ester 95%

Supplier: Apollo Scientific

Synonyms: 4,4,5,5-tetramethyl-2-(3-((tetrahydro-2H-pyran-2-yl)oxy)phenyl)-1,3,2-dioxaborolane

APOSOR4121-1GEA 126 EUR
APOSOR4121-1G APOSOR4121-5G
3-(2-Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, pinacol ester 95%
3-(2-Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, pinacol ester
Formula: C₁₇H₂₅BO₄
MW: 304,189 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659834
CAS Number: 850568-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR