4-(Thiomorpholin-4-ylsulphonyl)benzeneboronic acid 98%

Supplier: Apollo Scientific

Synonyms: N-Thiomorpholin-4-yl 4-boronobenzenesulphonamide

APOSOR4124-1GEA 483 EUR
APOSOR4124-1G APOSOR4124-5G
4-(Thiomorpholin-4-ylsulphonyl)benzeneboronic acid 98%
4-(Thiomorpholin-4-ylsulphonyl)benzeneboronic acid
Formula: C₁₀H₁₄BNO₄S₂
MW: 287,163 g/mol
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD07363757
CAS Number: 871329-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR