4-(1,3,4-Oxadiazol-2-yl)benzaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 2-(4-Formylphenyl)-1,3,4-oxadiazole

APOSOR42225-1GEA 205 EUR
APOSOR42225-1G
4-(1,3,4-Oxadiazol-2-yl)benzaldehyde
4-(1,3,4-Oxadiazol-2-yl)benzaldehyde
Formula: C₉H₆N₂O₂
MW: 174,156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18909697
CAS Number: 545424-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR