3-[3-(Morpholin-4-yl)propyl]benzeneboronic acid, pinacol ester hydrochloride 98%

Supplier: Apollo Scientific

Synonyms: 4-{3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propyl}morpholine hydrochloride

APOSOR43574-250MGEA 553 EUR
APOSOR43574-250MG
3-[3-(Morpholin-4-yl)propyl]benzeneboronic acid, pinacol ester hydrochloride 98%
3-[3-(Morpholin-4-yl)propyl]benzeneboronic acid, pinacol ester hydrochloride
Formula: C₁₉H₃₀BNO₃·ClH
MW: 367,718 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026080
CAS Number: 1150271-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR