(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Hydroxymethyl)-1H-pyrrolo[3,2-b]pyridine
APOSOR43672-1GEA
531
EUR
APOSOR43672-1G
(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol
(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol
Formula:
C₈H₈N₂O MW: 148,162 g/mol Melting Pt: 198…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD18432555 CAS Number: 17288-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...