3-(Pyridin-3-yl)aniline 97%
|
Warning
|
Synonyms:
3-(Pyridin-3-yl)benzenamine
APOSOR4377-250MGEA
0
EUR
APOSOR4377-250MG
3-(Pyridin-3-yl)aniline 97%
3-(Pyridin-3-yl)aniline
Formula:
C₁₁H₁₀N₂ MW: 170,211 g/mol Melting Pt: 72…74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07368549 CAS Number: 57976-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...