N,N-Dimethyl-2-azetidinecarboxamide hydrochloride
Supplier: Apollo Scientific
APOSOR451007-1GEA
1160
EUR
APOSOR451007-1G
N,N-Dimethyl-2-azetidinecarboxamide hydrochloride
N,N-Dimethyl-2-azetidinecarboxamide hydrochloride
Formula:
C₆H₁₂N₂O·ClH MW: 164,633 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28895101 CAS Number: 787563-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...