Methyl 1-(Methylsulphonyl)-3-azetidinecarboxylate

Supplier: Apollo Scientific

APOSOR451040-1GEA 1160 EUR
APOSOR451040-1G
Methyl 1-(Methylsulphonyl)-3-azetidinecarboxylate
Methyl 1-(Methylsulphonyl)-3-azetidinecarboxylate
Formula: C₆H₁₁NO₄S
MW: 193,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22682788
CAS Number: 1418117-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR