Ethyl 3-cyclopentyl-3-oxopropionate

Supplier: Apollo Scientific

Synonyms: Ethyl 3-cyclopentyl-3-oxopropanoate

APOSOR451200-25GEA 978 EUR
APOSOR451200-25G
Ethyl 3-cyclopentyl-3-oxopropionate
Ethyl 3-cyclopentyl-3-oxopropionate
Formula: C₁₀H₁₆O₃
MW: 184,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410650
CAS Number: 24922-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR