1-Boc-4-(4-piperidinyl)-piperazine 97%
Supplier: Apollo Scientific
Synonyms:
tert-Butyl 4-(piperidin-4-yl)piperazine-1-carboxylate
APOSOR452111-1GEA
394
EUR
APOSOR452111-1G
APOSOR452111-500MG
1-Boc-4-(4-piperidinyl)-piperazine 97%
1-Boc-4-(4-piperidinyl)-piperazine
Formula:
C₁₄H₂₇N₃O₂ MW: 269,383 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04974009 CAS Number: 205059-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...