endo-N,9-Dimethyl-9-azabicyclo[3.3.1]nonan-3-amine
Supplier: Apollo Scientific
APOSOR470007-5GEA
1690
EUR
APOSOR470007-5G
endo-N,9-Dimethyl-9-azabicyclo[3.3.1]nonan-3-amine
endo-N,9-Dimethyl-9-azabicyclo[3.3.1]nonan-3-amine
Formula:
C₁₀H₂₀N₂ MW: 168,279 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12198905 CAS Number: 123259-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...