2-(2-Nitrophenyl)imidazole

Supplier: Apollo Scientific

APOSOR470196-1GEA 397 EUR
APOSOR470196-1G
2-(2-Nitrophenyl)imidazole
2-(2-Nitrophenyl)imidazole
Formula: C₉H₇N₃O₂
MW: 189,171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863372
CAS Number: 4205-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR