N-(4-Tetrahydropyranylmethyl)benzylamine

Supplier: Apollo Scientific

APOSOR470348-5GEA 1120 EUR
APOSOR470348-5G
N-(4-Tetrahydropyranylmethyl)benzylamine
N-(4-Tetrahydropyranylmethyl)benzylamine
Formula: C₁₃H₁₉NO
MW: 205,296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371441
CAS Number: 887978-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR