5-(2-Methylphenyl)-2-thiophenecarbaldehyde

Supplier: Apollo Scientific

APOSOR470356-5GEA 1540 EUR
APOSOR470356-5G
5-(2-Methylphenyl)-2-thiophenecarbaldehyde
5-(2-Methylphenyl)-2-thiophenecarbaldehyde
Formula: C₁₂H₁₀OS
MW: 202,272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410187
CAS Number: 886509-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR