2-Amino-1,3,5-triazino[1,2-a]benzimidazol-4(3H)-one
Supplier: Apollo Scientific
APOSOR470439-5GEA
804
EUR
APOSOR470439-5G
2-Amino-1,3,5-triazino[1,2-a]benzimidazol-4(3H)-one
2-Amino-1,3,5-triazino[1,2-a]benzimidazol-4(3H)-one
Formula:
C₉H₇N₅O MW: 201,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02252785 CAS Number: 52066-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...