5-(tert-Butyl)-2-ethylbenzoxazole

Supplier: Apollo Scientific

APOSOR470462-5GEA 408 EUR
APOSOR470462-5G
5-(tert-Butyl)-2-ethylbenzoxazole
5-(tert-Butyl)-2-ethylbenzoxazole
Formula: C₁₃H₁₇NO
MW: 203,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19375298
CAS Number: 1267427-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR