Ethyl 5-(4-Pyridinyl)pyrazole-3-carboxylate

Supplier: Apollo Scientific

APOSOR470512-5GEA 916 EUR
APOSOR470512-5G
Ethyl 5-(4-Pyridinyl)pyrazole-3-carboxylate
Ethyl 5-(4-Pyridinyl)pyrazole-3-carboxylate
Formula: C₁₁H₁₁N₃O₂
MW: 217,224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783845
CAS Number: 911461-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR