1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Supplier: Apollo Scientific
APOSOR470525-5GEA
1010
EUR
APOSOR470525-5G
1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
1-(3-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Formula:
C₁₄H₁₆O₄ MW: 248,274 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20257290 CAS Number: 51414-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...