Methyl 1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Supplier: Apollo Scientific
APOSOR470529-5GEA
1010
EUR
APOSOR470529-5G
Methyl 1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Formula:
C₁₅H₁₈O₄ MW: 262,301 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22423130 CAS Number: 1384264-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...