Methyl 2-[(5-Bromo-2-pyridyl)oxy]acetate
Supplier: Apollo Scientific
APOSOR470575-5GEA
713
EUR
APOSOR470575-5G
Methyl 2-[(5-Bromo-2-pyridyl)oxy]acetate
Methyl 2-[(5-Bromo-2-pyridyl)oxy]acetate
Formula:
C₈H₈BrNO₃ MW: 246,058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13190760 CAS Number: 845890-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...