1-Acetyl-3-piperidineacetic acid
Supplier: Apollo Scientific
APOSOR470622-1GEA
499
EUR
APOSOR470622-1G
1-Acetyl-3-piperidineacetic acid
1-Acetyl-3-piperidineacetic acid
Formula:
C₉H₁₅NO₃ MW: 185,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14582888 CAS Number: 169253-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...