2-Benzyl-3-hydroxypropanenitrile

Supplier: Apollo Scientific

APOSOR470632-1GEA 321 EUR
APOSOR470632-1G APOSOR470632-5G
2-Benzyl-3-hydroxypropanenitrile
2-Benzyl-3-hydroxypropanenitrile
Formula: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18294353
CAS Number: 2601-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR