Ethyl 5-[(Diphenylmethylene)amino]-2-pyrimidinecarboxylate

Supplier: Apollo Scientific

APOSOR470643-1GEA 713 EUR
APOSOR470643-1G
Ethyl 5-[(Diphenylmethylene)amino]-2-pyrimidinecarboxylate
Ethyl 5-[(Diphenylmethylene)amino]-2-pyrimidinecarboxylate
Formula: C₂₀H₁₇N₃O₂
MW: 331,368 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24335252
CAS Number: 1434128-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR