1-Cbz-3-[Boc(cyclopropyl)amino]piperidine
Supplier: Apollo Scientific
APOSOR470678-5GEA
1210
EUR
APOSOR470678-5G
1-Cbz-3-[Boc(cyclopropyl)amino]piperidine
1-Cbz-3-[Boc(cyclopropyl)amino]piperidine
Formula:
C₂₁H₃₀N₂O₄ MW: 374,474 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26404523 CAS Number: 1823267-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...