5-(Hydroxymethyl)-2-(methylthio)pyrimidine

Supplier: Apollo Scientific

Synonyms: (2-(Methylthio)pyrimidin-5-yl)methanol

APOSOR470724-1GEA 232 EUR
APOSOR470724-1G
5-(Hydroxymethyl)-2-(methylthio)pyrimidine
5-(Hydroxymethyl)-2-(methylthio)pyrimidine
Formula: C₆H₈N₂OS
MW: 156,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053756
CAS Number: 19858-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR