tert-Butyl (S)-3-(Boc-amino)-4-iodobutanoate

Supplier: Apollo Scientific

APOSOR470803-1GEA 804 EUR
APOSOR470803-1G
tert-Butyl (S)-3-(Boc-amino)-4-iodobutanoate
tert-Butyl (S)-3-(Boc-amino)-4-iodobutanoate
Formula: C₁₃H₂₄INO₄
MW: 385,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27500655
CAS Number: 1802736-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR