4-Chloro-2-methylbutanoyl chloride
Supplier: Apollo Scientific
APOSOR470808-5GEA
1120
EUR
APOSOR470808-5G
4-Chloro-2-methylbutanoyl chloride
4-Chloro-2-methylbutanoyl chloride
Formula:
C₅H₈Cl₂O MW: 155,022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19232042 CAS Number: 51708-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...