2-Methoxy-7-methylnaphthalene

Supplier: Apollo Scientific

APOSOR470814-1GEA 300 EUR
APOSOR470814-1G
2-Methoxy-7-methylnaphthalene
2-Methoxy-7-methylnaphthalene
Formula: C₁₂H₁₂O
MW: 172,223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18412839
CAS Number: 1131-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR