Ethyl 2-Methyltetrahydrofuran-2-carboxylate
Supplier: Apollo Scientific
APOSOR470844-5GEA
916
EUR
APOSOR470844-5G
Ethyl 2-Methyltetrahydrofuran-2-carboxylate
Ethyl 2-Methyltetrahydrofuran-2-carboxylate
Formula:
C₈H₁₄O₃ MW: 158,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23847310 CAS Number: 1540337-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...