2-Bromo-5-iodothiophene
Supplier: Apollo Scientific
APOSOR470871-5GEA
713
EUR
APOSOR470871-5G
2-Bromo-5-iodothiophene
2-Bromo-5-iodothiophene
Formula:
C₄H₂BrIS MW: 288,932 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08695058 CAS Number: 29504-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...