1-Hydroxy-1-phenyl-2-propanone

Supplier: Apollo Scientific
Warning

APOSOR470879-250MGEA 219 EUR
APOSOR470879-250MG APOSOR470879-5G APOSOR470879-1G
1-Hydroxy-1-phenyl-2-propanone
1-Hydroxy-1-phenyl-2-propanone
Formula: C₉H₁₀O₂
MW: 150,175 g/mol
Boiling Pt: 124…125 °C (12 mm Hg)
Melting Pt: 44509 °C
Density: 1,105 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03792883
CAS Number: 90-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR