2,2-Dimethyl-2H-chromene-6-carbaldehyde
Supplier: Apollo Scientific
APOSOR470894-1GEA
804
EUR
APOSOR470894-1G
2,2-Dimethyl-2H-chromene-6-carbaldehyde
2,2-Dimethyl-2H-chromene-6-carbaldehyde
Formula:
C₁₂H₁₂O₂ MW: 188,222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17215738 CAS Number: 69964-40-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...