2-[3-(Methylsulphonyl)phenyl]ethylamine hydrochloride
Supplier: Apollo Scientific
APOSOR470915-5GEA
1010
EUR
APOSOR470915-5G
2-[3-(Methylsulphonyl)phenyl]ethylamine hydrochloride
2-[3-(Methylsulphonyl)phenyl]ethylamine hydrochloride
Formula:
C₉H₁₃NO₂S·ClH MW: 235,731 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27986815 CAS Number: 1073666-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...