N-Boc-2-(1-Iminoethyl)hydrazine
Supplier: Apollo Scientific
APOSOR470919-1GEA
888
EUR
APOSOR470919-1G
N-Boc-2-(1-Iminoethyl)hydrazine
N-Boc-2-(1-Iminoethyl)hydrazine
Formula:
C₇H₁₅N₃O₂ MW: 173,213 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27986814 CAS Number: 851535-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...