2,6-Dimethylpyridine-3-boronic acid pinacol ester

Supplier: Apollo Scientific

Synonyms: 2,6-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

APOSOR470942-1GEA 1090 EUR
APOSOR470942-1G
2,6-Dimethylpyridine-3-boronic acid pinacol ester
2,6-Dimethylpyridine-3-boronic acid pinacol ester
Formula: C₁₃H₂₀BNO₂
MW: 233,114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195066
CAS Number: 693774-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR