1-Cyclopropyl-1-(4-methoxyphenyl)methylamine

Supplier: Apollo Scientific

APOSOR471040-1GEA 553 EUR
APOSOR471040-1G
1-Cyclopropyl-1-(4-methoxyphenyl)methylamine
1-Cyclopropyl-1-(4-methoxyphenyl)methylamine
Formula: C₁₁H₁₅NO
MW: 177,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044830
CAS Number: 54398-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR