1-(2-Methoxyethyl)cyclopropanecarboxylic acid

Supplier: Apollo Scientific

APOSOR471191-1GEA 916 EUR
APOSOR471191-1G
1-(2-Methoxyethyl)cyclopropanecarboxylic acid
1-(2-Methoxyethyl)cyclopropanecarboxylic acid
Formula: C₇H₁₂O₃
MW: 144,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21669750
CAS Number: 1423031-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR