Ethyl 2-Oxo-2-[(2-oxo-2-phenylethyl)amino]acetate
Supplier: Apollo Scientific
APOSOR471209-1GEA
493
EUR
APOSOR471209-1G
Ethyl 2-Oxo-2-[(2-oxo-2-phenylethyl)amino]acetate
Ethyl 2-Oxo-2-[(2-oxo-2-phenylethyl)amino]acetate
Formula:
C₁₂H₁₃NO₄ MW: 235,236 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22493454 CAS Number: 84978-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...