5-(Aminomethyl)-1,4-benzodioxane

Supplier: Apollo Scientific

APOSOR471269-5GEA 1540 EUR
APOSOR471269-5G
5-(Aminomethyl)-1,4-benzodioxane
5-(Aminomethyl)-1,4-benzodioxane
Formula: C₉H₁₁NO₂
MW: 165,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972618
CAS Number: 261633-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR