Ethyl 2,4,6-Trichloronicotinate

Supplier: Apollo Scientific

APOSOR471384-1GEA 1010 EUR
APOSOR471384-1G
Ethyl 2,4,6-Trichloronicotinate
Ethyl 2,4,6-Trichloronicotinate
Formula: C₈H₆Cl₃NO₂
MW: 254,498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503376
CAS Number: 1934657-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR