Ethyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate

Supplier: Apollo Scientific

APOSOR471406-1GEA 1220 EUR
APOSOR471406-1G
Ethyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Ethyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Formula: C₁₃H₁₂ClNO₄
MW: 281,692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977384
CAS Number: 2006277-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR