(S)-3-Methyl-2-(1-pyrrolidinyl)butyric acid

Supplier: Apollo Scientific

APOSOR471423-1GEA 804 EUR
APOSOR471423-1G
(S)-3-Methyl-2-(1-pyrrolidinyl)butyric acid
(S)-3-Methyl-2-(1-pyrrolidinyl)butyric acid
Formula: C₉H₁₇NO₂
MW: 171,237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20656283
CAS Number: 777839-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR