1-Acetyl-3-(isopropylamino)piperidine
Supplier: Apollo Scientific
APOSOR471437-1GEA
1540
EUR
APOSOR471437-1G
1-Acetyl-3-(isopropylamino)piperidine
1-Acetyl-3-(isopropylamino)piperidine
Formula:
C₁₀H₂₀N₂O MW: 184,279 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21098064 CAS Number: 1353973-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...