4-(Benzyloxy)-3-hydroxyphenylacetic acid

Supplier: Apollo Scientific

APOSOR471563-1GEA 1010 EUR
APOSOR471563-1G
4-(Benzyloxy)-3-hydroxyphenylacetic acid
4-(Benzyloxy)-3-hydroxyphenylacetic acid
Formula: C₁₅H₁₄O₄
MW: 258,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16293773
CAS Number: 28988-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR